2-Fluor-4-pyridinboronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoropyridine-4-boronic acid pinacol ester, 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

AS50257G1 AS50257G5
ACTIAS50257G1EA 0 CHF
ACTIAS50257G1 ACTIAS50257G5
2-Fluor-4-pyridinboronsäurepinakolester
2-Fluor-4-pyridinboronsäurepinakolester
Formel: C₁₁H₁₅BFNO₂
MW: 223.05 g/mol
MDL Number: MFCD06798253
CAS-Nummer: 458532-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR