3-Brom-2-fluor-4-pyridinboronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Bromo-2-fluoropyridine-4-boronic acid pinacol ester

AS51312G1 AS51312G5
ACTIAS51312G1EA 0 CHF
ACTIAS51312G1 ACTIAS51312G5
3-Brom-2-fluor-4-pyridinboronsäurepinakolester
3-Brom-2-fluor-4-pyridinboronsäurepinakolester
Formel: C₁₁H₁₄BBrFNO₂
MW: 301.95 g/mol
MDL Number: MFCD08063110
CAS-Nummer: 1150561-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR