2-Brom-4-pyridinboronsäurepinakolester
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Bromo-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(2-Bromo-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Bromopyridine-4-boronic acid pinacol ester, 2-Bromo-4-pyridylboronic acid Pinacol ester
AS34395G1
AS34395G5
ACTIAS34395G1EA
0
CHF
ACTIAS34395G1
ACTIAS34395G5
2-Brom-4-pyridinboronsäurepinakolester
2-Brom-4-pyridinboronsäurepinakolester
Formel:
C₁₁H₁₅BBrNO₂ MW: 283.96 g/mol |
MDL Number:
MFCD06798252 CAS-Nummer: 458532-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...