5-Methylpyridin-3-ol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Hydroxy-5-methylpyridine, 3-Hydroxy-5-picoline

AS31287G1 AS31287G5
ACTIAS31287G1EA 50 CHF
ACTIAS31287G1 ACTIAS31287G5
5-Methylpyridin-3-ol
5-Methylpyridin-3-ol
Formel: C₆H₇NO
MW: 109,13 g/mol
MDL Number: MFCD00661297
CAS-Nummer: 42732-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR