Phenol : Chloroform : Isoamylalkohol (25:24:1) (v/v/v)

Lieferant: EUROCLONE
Gefahr

Synonyms: Carbolic acid, Hydroxybenzene, Phenic acid

EMR187100 EMR187500
EURLEMR187100EA 0 CHF
EURLEMR187100 EURLEMR187500
Phenol : Chloroform : Isoamylalkohol (25:24:1) (v/v/v)
Phenol : Chloroform : Isoamylalkohol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00133763
CAS-Nummer: 136112-00-0
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR