5-Methyl-2-(tributylstannyl)pyridin

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS51193G1 AS51193G5
ACTIAS51193G1EA 199 CHF
ACTIAS51193G1 ACTIAS51193G5
5-Methyl-2-(tributylstannyl)pyridin
5-Methyl-2-(tributylstannyl)pyridin
Formel: C₁₈H₃₃NSn
MW: 382,16 g/mol
MDL Number: MFCD07787402
CAS-Nummer: 189195-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR