5-Chlor-2-(tributylstannyl)pyridin

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS51733G1 AS51733G5
ACTIAS51733G1EA 0 CHF
ACTIAS51733G1 ACTIAS51733G5
5-Chlor-2-(tributylstannyl)pyridin
5-Chlor-2-(tributylstannyl)pyridin
Formel: C₁₇H₃₀ClNSn
MDL Number: MFCD10699156
CAS-Nummer: 611168-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR