2,4-Dichlor-6-methyl-3-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

AS30157G1 AS30157G5
ACTIAS30157G1EA 0 CHF
ACTIAS30157G1 ACTIAS30157G5
2,4-Dichlor-6-methyl-3-nitropyridin
2,4-Dichloro-6-methyl-3-nitropyridine
Formel: C₆H₄Cl₂N₂O₂
MDL Number: MFCD00218548
CAS-Nummer: 63897-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR