4-Chlor-2-picolin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Chloro-2-methylpyridine

AS31264G1 AS31264G5
ACTIAS31264G1EA 0 CHF
ACTIAS31264G1 ACTIAS31264G5
4-Chlor-2-picolin
4-Chlor-2-picolin
Formel: C₆H₆ClN
MW: 127.57 g/mol
MDL Number: MFCD00038833
CAS-Nummer: 3678-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR