2,6-Dichlor-3-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS36163G1 AS36163G5
ACTIAS36163G1EA 0 CHF
ACTIAS36163G1 ACTIAS36163G5
2,6-Dichlor-3-methylpyridin
2,6-Dichloro-3-methylpyridine
Formel: C₆H₅Cl₂N
MDL Number: MFCD00137847
CAS-Nummer: 58584-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR