2,4-Dichlor-5-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,4-Dichloro-5-picoline

AS34151G1 AS34151G5
ACTIAS34151G1EA 0 CHF
ACTIAS34151G1 ACTIAS34151G5
2,4-Dichlor-5-methylpyridin
2,4-Dichloro-5-methylpyridine
Formel: C₆H₅Cl₂N
MW: 162,02 g/mol
MDL Number: MFCD08704813
CAS-Nummer: 56961-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR