2,3-Dichlor-6-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS34498G1 AS34498G5
ACTIAS34498G1EA 0 CHF
ACTIAS34498G1 ACTIAS34498G5
2,3-Dichlor-6-methylpyridin
2,3-Dichloro-6-methylpyridine
Formel: C₆H₅Cl₂N
MW: 162.0166 g/mol
MDL Number: MFCD09839232
CAS-Nummer: 54957-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR