2,4-Dibrom-3-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

AS1952G1 AS1952G5
ACTIAS1952G1EA 0 CHF
ACTIAS1952G1 ACTIAS1952G5
2,4-Dibrom-3-nitropyridin
2,4-Dibromo-3-nitropyridine
Formel: C₅H₂Br₂N₂O₂
MDL Number: MFCD09750299
CAS-Nummer: 121263-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR