2,3-Dibrom-5-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

AS35168G1 AS35168G5
ACTIAS35168G1EA 0 CHF
ACTIAS35168G1 ACTIAS35168G5
2,3-Dibrom-5-nitropyridin
2,3-Dibromo-5-nitropyridine
Formel: C₅H₂Br₂N₂O₂
MW: 281,8896 g/mol
MDL Number: MFCD00233994
CAS-Nummer: 15862-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR