Ethyl-5-methylpicolinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl 5-methylpicolinate, Ethyl 5-methylpyridine-2-carboxylate

AS36389G1 AS36389G5
ACTIAS36389G1EA 0 CHF
ACTIAS36389G1 ACTIAS36389G5
Ethyl-5-methylpicolinat
Ethyl-5-methylpicolinat
Formel: C₉H₁₁NO₂
MW: 165,19 g/mol
MDL Number: MFCD10566273
CAS-Nummer: 55876-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR