Methyl-3-methylpicolinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 3-Methylpicolinate, Methyl 3-Methylpyridine-2-carboxylate, Methyl 3-methylpicolinate, 3-Methylpyridine-2-carboxylic acid methyll ester, 3-Methylpicolinic acid methyll ester

AS21748G25 AS21748G5
ACTIAS21748G25EA 0 CHF
ACTIAS21748G25 ACTIAS21748G5
Methyl-3-methylpicolinat
Methyl-3-methylpicolinat
Formel: C₈H₉NO₂
MW: 151.1600 g/mol
MDL Number: MFCD03789583
CAS-Nummer: 59718-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR