Methyl-4-methoxypicolinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 4-methoxypyridine-2-carboxylate

AS35527G1 AS35527G5
ACTIAS35527G1EA 0 CHF
ACTIAS35527G1 ACTIAS35527G5
Methyl-4-methoxypicolinat
Methyl-4-methoxypicolinat
Formel: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD00956223
CAS-Nummer: 29681-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR