Methyl-5-aminopicolinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 5-aminopyridine-2-carboxylate, Methyl 5-aminopicolinate

AS30885G1 AS30885G5
ACTIAS30885G1EA 36 CHF
ACTIAS30885G1 ACTIAS30885G5
Methyl-5-aminopicolinat
Methyl-5-aminopicolinat
Formel: C₇H₈N₂O₂
MW: 152,1506 g/mol
MDL Number: MFCD07783939
CAS-Nummer: 67515-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR