Methyl-6-chlorpicolinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 6-chloropyridine-2-carboxylate, Methyl 6-chloropicolinate

AS22729G25 AS22729G5
ACTIAS22729G25EA 54 CHF
ACTIAS22729G25 ACTIAS22729G5
Methyl-6-chlorpicolinat
Methyl-6-chlorpicolinat
Formel: C₇H₆ClNO₂
MW: 171,58 g/mol
MDL Number: MFCD00155391
CAS-Nummer: 6636-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR