Methyl-4,6-dichlorpicolinat

Lieferant: ACTIVATE SCIENTIFIC

AS33252G1 AS33252G5
ACTIAS33252G1EA 0 CHF
ACTIAS33252G1 ACTIAS33252G5
Methyl-4,6-dichlorpicolinat
Methyl-4,6-dichlorpicolinat
Formel: C₇H₅Cl₂NO₂
MDL Number: MFCD12965121
CAS-Nummer: 98273-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR