Methyl-3-chlorpicolinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 3-chloropicolinate, Methyl 3-chloropyridine-2-carboxylate

AS30642G1 AS30642G5
ACTIAS30642G1EA 0 CHF
ACTIAS30642G1 ACTIAS30642G5
Methyl-3-chlorpicolinat
Methyl-3-chlorpicolinat
Formel: C₇H₆ClNO₂
MW: 171,5810 g/mol
MDL Number: MFCD09750958
CAS-Nummer: 116383-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR