Methyl-4-brompicolinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 4-bromopicolinate, Methyl 4-bromopyridine-2-carboxylate

AS13020G1 AS13020G5
ACTIAS13020G1EA 0 CHF
ACTIAS13020G1 ACTIAS13020G5
Methyl-4-brompicolinat
Methyl-4-brompicolinat
Formel: C₇H₆BrNO₂
MW: 216,03 g/mol
MDL Number: MFCD07374897
CAS-Nummer: 29681-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR