6-(Trifluormethyl)-4-chinolinol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Hydroxy-6-(trifluoromethyl)quinoline
AS20945G25
AS20945G5
ACTIAS20945G25EA
0
CHF
ACTIAS20945G25
ACTIAS20945G5
6-(Trifluormethyl)-4-chinolinol
6-(Trifluormethyl)-4-chinolinol
Formel:
C₁₀H₆F₃NO MW: 213.16 g/mol |
MDL Number:
MFCD00209673 CAS-Nummer: 49713-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...