2-(Trifluormethyl)-4-chinolinol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-(Trifluoromethyl)quinolin-4-ol, 4-Hydroxy-2-(trifluoromethyl)quinoline

AS22147G1 AS22147G5
ACTIAS22147G1EA 0 CHF
ACTIAS22147G1 ACTIAS22147G5
2-(Trifluormethyl)-4-chinolinol
2-(Trifluormethyl)-4-chinolinol
Formel: C₁₀H₆F₃NO
MW: 213.1560 g/mol
MDL Number: MFCD00153196
CAS-Nummer: 1701-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR