Ethyl-5-chlor-2-thiophencarboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl 5-chlorothiophene-2-carboxylate

AS32417G1 AS32417G5
ACTIAS32417G1EA 0 CHF
ACTIAS32417G1 ACTIAS32417G5
Ethyl-5-chlor-2-thiophencarboxylat
Ethyl-5-chlor-2-thiophencarboxylat
Formel: C₇H₇ClO₂S
MW: 190,65 g/mol
MDL Number: MFCD00051732
CAS-Nummer: 5751-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR