3-(Dimethylamino)benzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS25494G1 AS25494G5
ACTIAS25494G1EA 110 CHF
ACTIAS25494G1 ACTIAS25494G5
3-(Dimethylamino)benzonitril
3-(Dimethylamino)benzonitril
Formel: C₉H₁₀N₂
MW: 146,1900 g/mol
MDL Number: MFCD00159008
CAS-Nummer: 38803-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR