2-Chlor-3-methoxyphenol
Lieferant: ACTIVATE SCIENTIFIC
AS30398G1
AS30398G5
ACTIAS30398G1EA
0
CHF
ACTIAS30398G1
ACTIAS30398G5
2-Chlor-3-methoxyphenol
2-Chlor-3-methoxyphenol
Formel:
C₇H₇ClO₂ MW: 158,5823 g/mol |
MDL Number:
MFCD08275690 CAS-Nummer: 72232-49-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...