4-Chlor-2,6-dinitrobenzoesäure
Lieferant: ACTIVATE SCIENTIFIC
AS23900G1
AS23900G5
ACTIAS23900G1EA
0
CHF
ACTIAS23900G1
ACTIAS23900G5
4-Chlor-2,6-dinitrobenzoesäure
4-Chloro-2,6-dinitrobenzoic acid
Formel:
C₇H₃ClN₂O₆ |
MDL Number:
MFCD02249880 CAS-Nummer: 95192-57-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...