2-Chlor-3-fluor-6-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS33540G1 AS33540G5
ACTIAS33540G1EA 0 CHF
ACTIAS33540G1 ACTIAS33540G5
2-Chlor-3-fluor-6-methylpyridin
2-Chloro-3-fluoro-6-methylpyridine
Formel: C₆H₅ClFN
MW: 145,56 g/mol
MDL Number: MFCD03095216
CAS-Nummer: 374633-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR