2-Brom-5-fluor-3-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS32525G1 AS32525G5
ACTIAS32525G1EA 0 CHF
ACTIAS32525G1 ACTIAS32525G5
2-Brom-5-fluor-3-methylpyridin
2-Bromo-5-fluoro-3-methylpyridine
Formel: C₆H₅BrFN
MDL Number: MFCD09835355
CAS-Nummer: 38186-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR