3-Fluor-4-methoxyphenol

Lieferant: ACTIVATE SCIENTIFIC

AS35757G1 AS35757G5
ACTIAS35757G1EA 0 CHF
ACTIAS35757G1 ACTIAS35757G5
3-Fluor-4-methoxyphenol
3-Fluor-4-methoxyphenol
Formel: C₇H₇FO₂
MDL Number: MFCD09263471
CAS-Nummer: 452-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR