3-Fluor-5-methoxyphenol

Lieferant: ACTIVATE SCIENTIFIC

AS34474G1 AS34474G5
ACTIAS34474G1EA 0 CHF
ACTIAS34474G1 ACTIAS34474G5
3-Fluor-5-methoxyphenol
3-Fluor-5-methoxyphenol
Formel: C₇H₇FO₂
MDL Number: MFCD11848782
CAS-Nummer: 850793-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR