5-Amino-2-fluor-3-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Amino-2-fluoro-3-picoline, 6-Fluoro-5-methylpyridin-3-amine

AS33413G1 AS33413G10 AS33413G5
ACTIAS33413G1EA 0 CHF
ACTIAS33413G1 ACTIAS33413G10 ACTIAS33413G5
5-Amino-2-fluor-3-methylpyridin
5-Amino-2-fluor-3-methylpyridin
Formel: C₆H₇FN₂
MW: 126.13 g/mol
MDL Number: MFCD03095088
CAS-Nummer: 186593-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR