2-Amino-3-chlor-5-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

AS30385G1 AS30385G5
ACTIAS30385G1EA 0 CHF
ACTIAS30385G1 ACTIAS30385G5
2-Amino-3-chlor-5-nitropyridin
2-Amino-3-chloro-5-nitropyridine
Formel: C₅H₄ClN₃O₂
MW: 173,5600 g/mol
MDL Number: MFCD00160299
CAS-Nummer: 22353-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR