3-Amino-2-chlor-6-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Amino-2-chloro-6-picoline, 5-Amino-6-chloro-2-picoline

AS31587G1 AS31587G5
ACTIAS31587G1EA 0 CHF
ACTIAS31587G1 ACTIAS31587G5
3-Amino-2-chlor-6-methylpyridin
3-Amino-2-chloro-6-methylpyridine
Formel: C₆H₇ClN₂
MW: 142,5900 g/mol
MDL Number: MFCD03095213
CAS-Nummer: 39745-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR