2-Amino-6-chlor-4-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Chloro-4-methylpyridin-2-amine

AS33626G1 AS33626G5
ACTIAS33626G1EA 0 CHF
ACTIAS33626G1 ACTIAS33626G5
2-Amino-6-chlor-4-methylpyridin
2-Amino-6-chloro-4-methylpyridine
Formel: C₆H₇ClN₂
MW: 142,5862 g/mol
MDL Number: MFCD09839256
CAS-Nummer: 51564-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR