2-Amino-5-chlor-4-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS7166G1 AS7166G5
ACTIAS7166G1EA 0 CHF
ACTIAS7166G1 ACTIAS7166G5
2-Amino-5-chlor-4-methylpyridin
2-Amino-5-chloro-4-methylpyridine
Formel: C₆H₇ClN₂
MDL Number: MFCD06410759
CAS-Nummer: 36936-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR