5-Amino-2-brom-3-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Amino-2-bromo-3-picoline

AS31268G10 AS31268G5
ACTIAS31268G10EA 0 CHF
ACTIAS31268G10 ACTIAS31268G5
5-Amino-2-brom-3-methylpyridin
5-Amino-2-brom-3-methylpyridin
Formel: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD03095085
CAS-Nummer: 38186-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR