2-Methoxy-6-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS24817G100 AS24817G25
ACTIAS24817G100EA 416 CHF
ACTIAS24817G100 ACTIAS24817G25
2-Methoxy-6-methylpyridin
2-Methoxy-6-methylpyridine
Formel: C₇H₉NO
MW: 123,16 g/mol
MDL Number: MFCD04112507
CAS-Nummer: 63071-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR