2-Methoxy-5-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS35211G1 AS35211G10 AS35211G5
ACTIAS35211G1EA 0 CHF
ACTIAS35211G1 ACTIAS35211G10 ACTIAS35211G5
2-Methoxy-5-methylpyridin
2-Methoxy-5-methylpyridine
Formel: C₇H₉NO
MW: 123,1500 g/mol
MDL Number: MFCD08703669
CAS-Nummer: 13472-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR