6-Methoxy-2-methyl-3-nitropyridin
Lieferant: ACTIVATE SCIENTIFIC
AS35396G1
AS35396G5
ACTIAS35396G1EA
0
CHF
ACTIAS35396G1
ACTIAS35396G5
6-Methoxy-2-methyl-3-nitropyridin
6-Methoxy-2-methyl-3-nitropyridin
Formel:
C₇H₈N₂O₃ MW: 168,1500 g/mol |
MDL Number:
MFCD03095076 CAS-Nummer: 5467-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...