2-Methoxy-4-methyl-3-nitropyridin
Lieferant: ACTIVATE SCIENTIFIC
AS36079G1
AS36079G5
ACTIAS36079G1EA
0
CHF
ACTIAS36079G1
ACTIAS36079G5
2-Methoxy-4-methyl-3-nitropyridin
2-Methoxy-4-methyl-3-nitropyridine
Formel:
C₇H₈N₂O₃ |
MDL Number:
MFCD08277286 CAS-Nummer: 160590-36-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...