2-Chlor-5-formyl-4-picolin

Lieferant: ACTIVATE SCIENTIFIC

AS32890G1 AS32890G5
ACTIAS32890G1EA 0 CHF
ACTIAS32890G1 ACTIAS32890G5
2-Chlor-5-formyl-4-picolin
2-Chlor-5-formyl-4-picolin
Formel: C₇H₆ClNO
MDL Number: MFCD07368881
CAS-Nummer: 884495-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR