4-Formyl-3-picolin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Methylnicotinaldehyde

AS12516G1 AS12516G5
ACTIAS12516G1EA 0 CHF
ACTIAS12516G1 ACTIAS12516G5
4-Formyl-3-picolin
4-Formyl-3-picolin
Formel: C₇H₇NO
MDL Number: MFCD07437997
CAS-Nummer: 51227-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR