4-Formyl-2-picolin

Lieferant: ACTIVATE SCIENTIFIC

AS36470G1 AS36470G5
ACTIAS36470G1EA 0 CHF
ACTIAS36470G1 ACTIAS36470G5
4-Formyl-2-picolin
4-Formyl-2-picolin
Formel: C₇H₇NO
MDL Number: MFCD06410683
CAS-Nummer: 63875-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR