Methyl-6-(brommethyl)picolinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 6-(bromomethyl)picolinate

AS36469G1 AS36469G5
ACTIAS36469G1EA 0 CHF
ACTIAS36469G1 ACTIAS36469G5
Methyl-6-(brommethyl)picolinat
Methyl-6-(brommethyl)picolinat
Formel: C₈H₈BrNO₂
MW: 230,0600 g/mol
MDL Number: MFCD03265368
CAS-Nummer: 146462-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR