Methyl-6-cyanpicolinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Cyano-2-pyridine carboxylic acid methyl ester, Methyl 6-cyanopicolinate

AS36693G1 AS36693G5
ACTIAS36693G1EA 0 CHF
ACTIAS36693G1 ACTIAS36693G5
Methyl-6-cyanpicolinat
Methyl-6-cyanpicolinat
Formel: C₈H₆N₂O₂
MW: 162,1500 g/mol
MDL Number: MFCD07367924
CAS-Nummer: 98436-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR