Methyl-4-isochinolincarboxylat

Lieferant: ACTIVATE SCIENTIFIC

AS36467G1 AS36467G5
ACTIAS36467G1EA 0 CHF
ACTIAS36467G1 ACTIAS36467G5
Methyl-4-isochinolincarboxylat
Methyl-4-isochinolincarboxylat
Formel: C₁₁H₉NO₂
MDL Number: MFCD11111833
CAS-Nummer: 20317-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR