Methyl-3-isochinolincarboxylat

Lieferant: ACTIVATE SCIENTIFIC

AS35104G1 AS35104G10 AS35104G5
ACTIAS35104G1EA 0 CHF
ACTIAS35104G1 ACTIAS35104G10 ACTIAS35104G5
Methyl-3-isochinolincarboxylat
Methyl-3-isochinolincarboxylat
Formel: C₁₁H₉NO₂
MDL Number: MFCD00075138
CAS-Nummer: 27104-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR