Ethyl-3-isochinolincarboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl 3-isoquinolinecarboxylate

AS30905G1 AS30905G5
ACTIAS30905G1EA 0 CHF
ACTIAS30905G1 ACTIAS30905G5
Ethyl-3-isochinolincarboxylat
Ethyl-3-isochinolincarboxylat
Formel: C₁₂H₁₁NO₂
MW: 201.2200 g/mol
MDL Number: MFCD10566077
CAS-Nummer: 50458-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR